Merck
  • Pseudomonas aeruginosa quorum-sensing signaling molecule N-3-oxododecanoyl homoserine lactone induces matrix metalloproteinase 9 expression via the AP1 pathway in rat fibroblasts.
Pseudomonas aeruginosa quorum-sensing signaling molecule N-3-oxododecanoyl homoserine lactone induces matrix metalloproteinase 9 expression via the AP1 pathway in rat fibroblasts.
Bioscience, biotechnology, and biochemistry (2015-06-23)
Gojiro Nakagami, Takeo Minematsu, Tomohiro Morohoshi, Takumi Yamane, Toshiki Kanazawa, Lijuan Huang, Mayumi Asada, Takashi Nagase, Shin-ichi Ikeda, Tsukasa Ikeda, Hiromi Sanada
ZUSAMMENFASSUNG
Quorum sensing is a cell-to-cell communication mechanism, which is responsible for regulating a number of bacterial virulence factors and biofilm maturation and therefore plays an important role for establishing wound infection. Quorum-sensing signals may induce inflammation and predispose wounds to infection by Pseudomonas aeruginosa; however, the interaction has not been well investigated. We examined the effects of the P. aeruginosa las quorum-sensing signal, N-3-oxo-dodecanoyl homoserine lactone (3OC12-HSL), on matrix metalloproteinase (MMP) 9 expression in Rat-1 fibroblasts. 3OC12-HSL upregulated the expression of the MMP9 gene bearing an activator protein-1 (AP-1) binding site in the promoter region. We further investigated the mechanism underlying this effect. c-Fos gene expression increased rapidly after exposure to 3OC12-HSL, and nuclear translocation of c-Fos protein was observed; both effects were reduced by pretreatment with an AP-1 inhibitor. These results suggest that 3OC12-HSL can alter MMP9 gene expression in fibroblasts via the AP-1 signaling pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinon, ≥99.0% (GC)
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinon, absolute, over molecular sieve (H2O ≤0.04%), ≥99.5% (GC)
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinon, reagent grade
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human FOS