Merck
  • Structural characterization and antioxidant activity of a novel heteropolysaccharide from the submerged fermentation mycelia of Ganoderma capense.
Structural characterization and antioxidant activity of a novel heteropolysaccharide from the submerged fermentation mycelia of Ganoderma capense.
Carbohydrate polymers (2015-10-03)
Yating Huang, Naisheng Li, Jian-Bo Wan, Dezhi Zhang, Chunyan Yan
ZUSAMMENFASSUNG
A novel heteropolysaccharide (GCPB-2) with a molecular mass of 1.03×10(5)Da was isolated from the submerged fermentation culturing mycelia powder of Lingzhi, Ganoderma capense, by DEAE-52 cellulose, DEAE Sepharose CL-6B, and Sephadex G-75 column chromatography. Its chemical structure was characterized for the first time. The antioxidant activity was evaluated by two antioxidant assay methods. The result of this study introduced G. capense as a possible valuable source that helped to exhibit some antioxidant properties. The homogeneous polysaccharide was composed of xylose and arabinose in the molar ratio of 1:1, and showed a specific optical rotation of [α]D(25)=+161° (c 1.0, H2O). Monosaccharide analysis, partial acid hydrolysis, periodic acid oxidation, methylation analysis, gas chromatography-mass spectrometry (GC-MS), Fourier transform-infrared spectroscopy (FT-IR), and nuclear magnetic resonance (NMR) spectroscopy ((1)H, (13)C, HMQC and HMBC) were conducted to elucidate its structure. The backbone of GCPB-2 was composed of (1 → 4)-linked-β-D-xylopyranose residues which branched at O-3. The two branches consisted of (1 → 3)-linked-β-L-arabinopyranose terminated with β-D-xylopyranose residues, and (1 → 4)-linked-β-L-arabinopyranose terminated with β-D-arabinopyranose residues. In the in vitro antioxidant assay, GCPB-2 was found to possess 1-diphenyl-2-picryl-hydrazyl (DPPH) radical-scavenging activity with an EC50 value of 0.379 μM. The findings of this study indicated that GCPB-2 possesses the hydroxyl radical-scavenging activity, which provided an experimental evidence to support the G. capense as functional foods in some Asian countries. To understand better the bioactivity of GCPB-2, the antioxidant activity should be further investigated to find out its mechanism in future work.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Schwefelsäure, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumhydrid, 60 % dispersion in mineral oil
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Sigma-Aldrich
Natriumhydrid, dry, 90%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Schwefelsäure, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis