Merck
  • The PEDF Neuroprotective Domain Plus DHA Induces Corneal Nerve Regeneration After Experimental Surgery.
The PEDF Neuroprotective Domain Plus DHA Induces Corneal Nerve Regeneration After Experimental Surgery.
Investigative ophthalmology & visual science (2015-06-02)
Jiucheng He, M Soledad Cortina, Azucena Kakazu, Haydee E P Bazan
ZUSAMMENFASSUNG
To compare a 44-mer pigment epithelial-derived factor (PEDF) peptide with neurotrophic activity, and a 34-mer PEDF with antiangiogenic properties in association with docosahexaenoic acid (DHA) in corneal nerve regeneration after experimental surgery. A corneal stromal dissection was performed in rabbits. Treatment groups received topical 44-mer, 34-mer, or full PEDF plus DHA. Corneal sensitivity and Schirmer's test were performed weekly. Rabbits were euthanized at 2 and 4 days and 8 weeks. Two- and 4-day samples were stained for neutrophils and CD11b+ cells. Corneal nerves were stained with βIII tubulin and calcitonin gene-related peptide (CGRP) antibodies in specimens collected at 8 weeks. Subepithelial nerve plexus density was calculated. A PEDF-receptor (PEDF-R) was analyzed in rabbit corneal epithelial cells (RCEC) by Western blot and immunofluorescence. Infiltration of CD11b+cells and neutrophils was reduced by treatment with 44-mer PEDF+DHA. A 3-fold increase in subepithelial corneal nerves and CGRP-positive nerves was found in the 44-mer PEDF+DHA-treated group compared with the 34-mer PEDF+DHA- and vehicle-treated groups. There was a 75% recovery of corneal sensitivity by week 7, and Schirmer's test reached control values in the 44-mer PEDF+DHA-treated corneas at 7 weeks. A PEDF-R protein with homology to calcium-independent phospholipase A2ς was expressed in RCEC. The 44-mer PEDF+DHA, but not the 34-mer PEDF+DHA, promotes functional regeneration of damaged corneal nerves. Forty four-mer PEDF, by activating a corneal epithelial receptor, in conjunction with DHA could be a novel therapeutic agent for the treatment of neurotrophic keratitis and dry eye that develops as a result of corneal nerve damage.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
(L)-Dehydroascorbinsäure
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
2-Phenylindol, technical grade, 95%
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Dehydro-L(+)-ascorbinsäure Dimer, ≥80% (enzymatic)