Merck
Curcumin nanoemulsion for transdermal application: formulation and evaluation.
Drug development and industrial pharmacy (2014-02-08)
Heni Rachmawati, Dewa Ken Budiputra, Rachmat Mauludin
ZUSAMMENFASSUNG
The aim of this work is to develop a curcumin nanoemulsion for transdermal delivery. The incorporation of curcumin inside a nanoglobul should improve curcumin stability and permeability. A nanoemulsion was prepared by the self-nanoemulsification method, using an oil phase of glyceryl monooleate, Cremophor RH40 and polyethylene glycol 400. Evaluation of the nanoemulsion included analysis of particle size, polydispersity index, zeta potential, physical stability, Raman spectrum and morphology. In addition, the physical performance of the nanoemulsion in Viscolam AT 100P gel was studied. A modified vertical diffusion cell and shed snake skin of Python reticulatus were used to study the in vitro permeation of curcumin. A spontaneously formed stable nanoemulsion has a loading capacity of 350 mg curcumin/10 g of oil phase. The mean droplet diameter, polydispersity index and zeta potential of optimized nanoemulsion were 85.0 ± 1.5 nm, 0.18 ± 0.0 and -5.9 ± 0.3 mV, respectively. Curcumin in a nanoemulsion was more stable than unencapsulated curcumin. Furthermore, nanoemulsification significantly improved the permeation flux of curcumin from the hydrophilic matrix gel; the release kinetic of curcumin changed from zero order to a Higuchi release profile. Overall, the developed nanoemulsion system not only improved curcumin permeability but also protected the curcumin from chemical degradation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Triethylamin, ≥99.5%
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Triethanolamin, ≥99.0% (GC)
Sigma-Aldrich
Triethylamin, ≥99%
Sigma-Aldrich
Triethylamin, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamin, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Sigma-Aldrich
L-Lysin -monohydrochlorid, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Kaliumphosphat, Molecular Biology, ≥98.0%
Sigma-Aldrich
Triethanolamin, reagent grade, 98%
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Triethylamin, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Kaliumphosphat, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
L-Lysin -monohydrochlorid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Triethanolamin, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamin, ≥99.5%
Sigma-Aldrich
Kaliumphosphat, 99.99% trace metals basis
Sigma-Aldrich
Triethanolamin, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
Triethylamin, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethanolamin, puriss., meets analytical specification of NF, ≥99% (GC)
Sigma-Aldrich
L-Lysin -monohydrochlorid, BioUltra, ≥99.5% (AT)