Merck
  • Waterborne exposure to microcystin-LR causes thyroid hormone metabolism disturbances in juvenile Chinese rare minnow (Gobiocypris rarus).
Waterborne exposure to microcystin-LR causes thyroid hormone metabolism disturbances in juvenile Chinese rare minnow (Gobiocypris rarus).
Environmental toxicology and chemistry (2015-04-23)
Zidong Liu, Dapeng Li, Ying Wang, Wei Guo, Yu Gao, Rong Tang
ZUSAMMENFASSUNG
Microcystin-LR (MC-LR) has the potential to disturb thyroid hormone homeostasis, but little is known about the underlying mechanisms of MC-LR in fish. In the present study, juvenile Chinese rare minnows (Gobiocypris rarus) were exposed to various concentrations of MC-LR (0 µg/L, 50 µg/L, 100 µg/L, and 500 µg/L) for 7 d. The whole-body thyroid hormone content, the histology of thyroid follicle epithelial cells, the activities of hepatic iodothyronine deiodinases, and the transcription of selected genes associated with thyroid hormone synthesis, transport, and metabolism were analyzed. Following exposure to MC-LR, whole-body concentrations of both thyroxine (T4 ) and triiodothyronine (T3 ) were significantly decreased. The levels of messenger RNA for sodium/iodide symporter, transthyretin, thyroid hormone receptor-α, iodothyronine deiodinase2, and iodothyronine deiodinase3 were significantly down-regulated after exposure to 500 µg/L MC-LR. A significant decrease in ID2 activity was also observed in the 500-µg/L MC-LR exposure group. Moreover, hypertrophy of thyroid follicle epithelial cells was observed after exposure to MC-LR. The results indicate that acute MC-LR exposure has the potential to disturb the homeostasis of thyroid hormone metabolism, leading to a hypothyroidism state in the juvenile Chinese rare minnow.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Trichloressigsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloressigsäure -Lösung, 6.1 N
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethyl-3-aminobenzoat -methansulfonat, 98%
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Trichloressigsäure, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Trichloressigsäure, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloressigsäure, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloressigsäure, ≥99.0% (titration)
Sigma-Aldrich
Trichloressigsäure, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®