Merck
  • Single nucleotide polymorphism of toll-like receptor 4 (TLR4) is associated with juvenile spondyloarthritis in Croatian population.
Single nucleotide polymorphism of toll-like receptor 4 (TLR4) is associated with juvenile spondyloarthritis in Croatian population.
Clinical rheumatology (2015-04-30)
Marija Perica, Mandica Vidović, Lovro Lamot, Lana Tambić Bukovac, Sanja Kapitanović, Magdalena Perić, Jerko Barbić, Miroslav Harjaček
ZUSAMMENFASSUNG
Single nucleotide polymorphisms (SNP) of toll-like and NOD-like receptors have been associated with altered receptor activity and modified production of proinflammatory cytokines leading to a number of diseases. Our aim was to determine whether SNP of TLR2 (Arg753Gln), TLR4 (Asp299Gly, Thr399Ile), and NLRP3 (Q705K) influence susceptibility to juvenile spondyloarthrtis (jSpA) and juvenile idiopathic arthritis (JIA). After the DNA extraction, 26 patients with jSpA, 11 with oligoarticular, polyarticular, or systemic JIA, and 40 healthy controls were genotyped for Arg753Gln, Asp299Gly, Thr399Ile, and Q705K SNP using real-time PCR-SNP analysis. Statistically significant difference in genotype frequency for Thr399Ile SNP of TLR4 was observed in the jSpA (χ2 = 6.705, p = 0.035) and not in the JIA group (χ2 = 3005, p = 0.223). Regarding Asp299Gly SNP, no significant difference in genotype frequency was found; however, allele frequency was significant in both jSpA and JIA patients. No significant difference in genotype or allele frequency was observed for Arg735Gln and Q705K SNP. The399Ile polymorphism of TLR4 may be responsible for altered immune response to microbial infection in variant carriers and represent a mechanism of triggering overproduction of proinflammatory cytokines and long-term inflammation in jSpA. SNP of TLR2, NLRP3, and TLR4 (Asp299Gly) were not associated with jSpA or JIA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri