Merck
  • Surfactant-controlled composition and crystal structure of manganese(II) sulfide nanocrystals prepared by solvothermal synthesis.
Surfactant-controlled composition and crystal structure of manganese(II) sulfide nanocrystals prepared by solvothermal synthesis.
Beilstein journal of nanotechnology (2016-01-07)
Elena Capetti, Anna M Ferretti, Vladimiro Dal Santo, Alessandro Ponti
ZUSAMMENFASSUNG
We investigated how the outcome of the solvothermal synthesis of manganese(II) sulfide (MnS) nanocrystals (NCs) is affected by the type and amount of long chain surfactant present in the reaction mixture. Prompted by a previous observation that a larger than stoichiometric amount of sulfur is required [Puglisi, A.; Mondini, S.; Cenedese, S.; Ferretti, A. M.; Santo, N.; Ponti A. Chem. Mater. 2010, 22, 2804-2813], we carried out a wide set of reactions using Mn(II) carboxylates and Mn2(CO)10 as precursors with varying amounts of sulfur and carboxylic acid. MnS NCs were obtained provided that the S/Mn ratio was larger than the L/Mn ratio, otherwise MnO NCs were produced. Since MnS can crystallize in three distinct phases (rock salt α-MnS, zincblende β-MnS, and wurtzite γ-MnS), we also investigated whether the surfactant affected the NC polymorphism. We found that MnS polymorphism can be controlled by appropriate selection of the surfactant. γ-MnS nanocrystals formed when a 1:2 mixture of long chain carboxylic acid and amine was used, irrespective of the presence of carboxylic acid as a free surfactant or ligand in the metal precursor. When we used a single surfactant (carboxylic acid, alcohol, thiol, amine), α-MnS nanocrystals were obtained. The peculiar role of the amine seems to be related to its basicity. The nanocrystals were characterized by TEM and electron diffraction; ATR-FTIR spectroscopy provided information about the surfactants adsorbed on the NCs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oleylamin, technical grade, 70%
Sigma-Aldrich
1-Octadecen, technical grade, 90%
Sigma-Aldrich
Oleinsäure, technical grade, 90%
Sigma-Aldrich
Tetramethylammoniumhydroxid Pentahydrat, ≥97%
Sigma-Aldrich
Oleinsäure, ≥99% (GC)
Sigma-Aldrich
1-Dodecanthiol, ≥98%
Sigma-Aldrich
Stearinsäure, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Oleinsäure, suitable for cell culture, BioReagent
Sigma-Aldrich
Schwefel, powder, 99.98% trace metals basis
Sigma-Aldrich
Schwefel, 99.998% trace metals basis
Sigma-Aldrich
Mangan(II)-chlorid, powder and chunks, ≥99% trace metals basis
Sigma-Aldrich
Stearinsäure, reagent grade, 95%
Sigma-Aldrich
Tetramethylammoniumhydroxid Pentahydrat, ≥95.0% (T)
Sigma-Aldrich
Oleinsäure, natural, FCC
Sigma-Aldrich
Oleinsäure, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
SAFC
Oleylamin, ≥98% (primary amine)
Sigma-Aldrich
Stearinsäure, ≥95%, FCC, FG
Sigma-Aldrich
Hexadecylamin, 98%
Sigma-Aldrich
Octadecylamin, ≥99% (GC)
Sigma-Aldrich
Dodecylamin, ≥99%
Sigma-Aldrich
Oleylalkohol, ≥99% (GC)
Sigma-Aldrich
Dodecylamin, 98%
Sigma-Aldrich
Mangan(II)-chlorid, flakes, 98%
Sigma-Aldrich
1-Octadecen, ≥95.0% (GC)
Sigma-Aldrich
Octadecylamin, ≥99.0% (GC)
Sigma-Aldrich
Oleylalkohol, technical grade, 85%
Sigma-Aldrich
Mangan(II)-chlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Hexadecylamin, technical grade, 90%
Sigma-Aldrich
Octadecylamin, technical grade, 90%
Sigma-Aldrich
Schwefel, flakes, ≥99.99% trace metals basis