Merck
  • Evaluation of (4-Arylpiperidin-1-yl)cyclopentanecarboxamides As High-Affinity and Long-Residence-Time Antagonists for the CCR2 Receptor.
Evaluation of (4-Arylpiperidin-1-yl)cyclopentanecarboxamides As High-Affinity and Long-Residence-Time Antagonists for the CCR2 Receptor.
ChemMedChem (2015-06-03)
Maris Vilums, Annelien J M Zweemer, Arian Dilanchian, Jacobus P D van Veldhoven, Henk de Vries, Johannes Brussee, John Saunders, Dean Stamos, Laura H Heitman, Adriaan P IJzerman
ZUSAMMENFASSUNG
Animal models suggest that the chemokine ligand 2/CC-chemokine receptor 2 (CCL2/CCR2) axis plays an important role in the development of inflammatory diseases. However, CCR2 antagonists have failed in clinical trials because of a lack of efficacy. We previously described a new approach for the design of CCR2 antagonists by the use of structure-kinetics relationships (SKRs). Herein we report new findings on the structure-affinity relationships (SARs) and SKRs of the reference compound MK-0483, its diastereomers, and its structural analogues as CCR2 antagonists. The SARs of the 4-arylpiperidine group suggest that lipophilic hydrogen-bond-accepting substituents at the 3-position are favorable. However, the SKRs suggest that a lipophilic group with a certain size is desired [e.g., 3-Br: Ki =2.8 nM, residence time (t(res))=243 min; 3-iPr: Ki =3.6 nM, t(res) =266 min]. Alternatively, additional substituents and further optimization of the molecule, while keeping a carboxylic acid at the 3-position, can also prolong t(res); this was most prominently observed in MK-0483 (Ki =1.2 nM, t(res) =724 min) and a close analogue (Ki =7.8 nM) with a short residence time.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%