Merck
  • The N-terminal segment of glyceraldehyde-3-phosphate dehydrogenase of Haemonchus contortus interacts with complements C1q and C3.
The N-terminal segment of glyceraldehyde-3-phosphate dehydrogenase of Haemonchus contortus interacts with complements C1q and C3.
Parasite immunology (2015-09-04)
G V Vedamurthy, S Sahoo, I K Devi, S Murugavel, P Joshi
ZUSAMMENFASSUNG
Haemonchus contortus, an economically important blood-sucking parasite of sheep and goats, survives the harsh host gut environment by secreting a number of proteins referred as excretory/secretory (ES) products. Glyceraldehyde-3-phosphate dehydrogenase (GAPDH), a glycolytic enzyme, is one of the components of H. contortus ES products. The parasite enzyme binds to complement C3 and inhibits its activity. In this study, the C3-binding activity of the parasite GAPDH was mapped to the N-terminal part of the enzyme by generating defined recombinant fragments of the protein. The N-terminal fragment also trapped complement C1q but not C5 and inhibited complement-mediated lysis of sensitized sheep erythrocytes. Competitive binding assay indicates different binding regions for C1q and C3 proteins. GAPDH stimulated proliferation of goat peripheral blood mononuclear cells in vitro and reacted with the sera from H. contortus-infected animals. However, the fragments of GAPDH did not stimulate cell proliferation nor reacted with the infected animal sera. Furthermore, denatured GAPDH failed to react with the infected animal sera in dot blot suggesting conformation-dependent epitope. These results demonstrate an elegant strategy of the parasite to completely shut down complement activation and identify GAPDH as a promising target for future therapeutic intervention.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Acrylamid, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
N,N′-Methylenbisacrylamid, powder, Molecular Biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
N,N′-Methylenbis(acrylamid), 99%
Sigma-Aldrich
o-Phenylendiamin, flaked, 99.5%
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Sigma-Aldrich
Trypanblau, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamid -Lösung, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Trypanblau, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
o-Phenylendiamin, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
N,N′-Methylenbisacrylamid -Lösung, suitable for electrophoresis, 2% in H2O
Sigma-Aldrich
o-Phenylendiamin, tablet, 20 mg substrate per tablet
Sigma-Aldrich
N,N′-Methylenbisacrylamid, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
o-Phenylendiamin, sublimed, ≥99%
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%