Merck
  • Chirality of Single-Handed Twisted Titania Tubular Nanoribbons Prepared Through Sol-gel Transcription.
Chirality of Single-Handed Twisted Titania Tubular Nanoribbons Prepared Through Sol-gel Transcription.
Chirality (2015-05-23)
Sibing Wang, Chuanyong Zhang, Yi Li, Baozong Li, Yonggang Yang
ZUSAMMENFASSUNG
Single-handed twisted titania tubular nanoribbons were prepared through sol-gel transcription using a pair of enantiomers. Handedness was controlled by that of the template. The obtained samples were characterized using field-emission electron microscopy, transmission electron microscopy, diffuse reflectance circular dichroism (DRCD), and X-ray diffraction. The DRCD spectra indicated that the titania nanotubes exhibit optical activity. Although the tubular structure was destroyed after being calcined at 700 °C for 2.0 h, DRCD signals were still identified. However, the DRCD signals disappeared after being calcined at 1000 °C for 2.0 h. The optical activity of titania was proposed to be due to chiral defects. Previous results showed that straight titania tubes could be used as asymmetric autocatalysts, indicating that titania exhibit chirality at the angstrom level. Herein, it was found that they also exhibit DRCD signals, indicating that there are no obvious relationships between morphology at the nano level and chirality at the angstrom level. The nanotube chirality should originate from the chiral defects on the nanotube inner surface. The Fourier transform infrared spectra indicated that the chirality of the titania was transferred from the gelators through the hydrogen bonding between N-H and Ti-OH.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.9 atom % D
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Titan(IV)isopropoxid, 97%
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.5 atom % D
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Titan(IV)isopropoxid, 99.999% trace metals basis
Sigma-Aldrich
Titan(IV)isopropoxid, ≥97.0%
Sigma-Aldrich
Salzsäure -Lösung, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Dimethylsulfoxid-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dodecandisäure, 99%
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
(±)-trans-1,2-Diaminocyclohexan, 99%
Sigma-Aldrich
Octadecylamin, ≥99% (GC)
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Dimethylsulfoxid-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethylsulfoxid-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Tetramethylsilan, ≥99.0% (GC)
Sigma-Aldrich
(1R,2R)-(−)-1,2-Diaminocyclohexan, 98%
Sigma-Aldrich
Octadecylamin, ≥99.0% (GC)
Sigma-Aldrich
Dimethylsulfoxid-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Titan(IV)isopropoxid, packaged for use in deposition systems
Sigma-Aldrich
Octadecylamin, technical grade, 90%
Sigma-Aldrich
Dimethylsulfoxid-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Tetramethylsilan, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Dimethylsulfoxid-d6, "100%", 99.96 atom % D