Merck
  • Comparison of Lipoprotein-Associated Phospholipase A2 and High Sensitive C-Reactive Protein as Determinants of Metabolic Syndrome in Subjects without Coronary Heart Disease: In Search of the Best Predictor.
Comparison of Lipoprotein-Associated Phospholipase A2 and High Sensitive C-Reactive Protein as Determinants of Metabolic Syndrome in Subjects without Coronary Heart Disease: In Search of the Best Predictor.
International journal of endocrinology (2015-06-20)
Mónica Acevedo, Paola Varleta, Verónica Kramer, Giovanna Valentino, Teresa Quiroga, Carolina Prieto, Jacqueline Parada, Marcela Adasme, Luisa Briones, Carlos Navarrete
ZUSAMMENFASSUNG
High sensitivity C-reactive protein (hsCRP) is a marker of metabolic syndrome (MS) and cardiovascular (CV) disease. Lipoprotein-associated phospholipase A2 (Lp-PLA2) also predicts CV disease. There are no reports comparing these markers as predictors of MS. Methods. Cross-sectional study comparing Lp-PLA2 and hsCRP as predictors of MS in asymptomatic subjects was carried out; 152 subjects without known atherosclerosis participated. Data were collected on demographics, cardiovascular risk factors, anthropometric and biochemical measurements, and hsCRP and Lp-PLA2 activity levels. A logistic regression analysis was performed with each biomarker and receiver operating characteristic (ROC) curves were constructed for MS. Results. Mean age was 46 ± 11 years, and 38% of the subjects had MS. Mean Lp-PLA2 activity was 185 ± 48 nmol/mL/min, and mean hsCRP was 2.1 ± 2.2 mg/L. Subjects with MS had significantly higher levels of Lp-PLA2 (P = 0.03) and hsCRP (P < 0.0001) than those without MS. ROC curves showed that both markers predicted MS. Conclusion. Lp-PLA2 and hsCRP are elevated in subjects with MS. Both biomarkers were independent and significant predictors for MS, emphasizing the role of inflammation in MS. Further research is necessary to determine if inflammation predicts a higher risk for CV events in MS subjects.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
Kreatinin, anhydrous, ≥98%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Nicotinsäure, ≥99.5% (HPLC)
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Nicotinsäure, ≥98%
Sigma-Aldrich
Nicotinsäure, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Nicotinsäure, meets USP testing specifications