Merck
  • A chemometric optimization of method for determination of nitrosamines in gastric juices by GC-MS.
A chemometric optimization of method for determination of nitrosamines in gastric juices by GC-MS.
Journal of pharmaceutical and biomedical analysis (2015-09-08)
Mehmet Akyüz, Şevket Ata, Erdal Dinç
ZUSAMMENFASSUNG
A chemometrically optimized isolation procedure combined with gas chromatography-mass spectrometry detection technique has been proposed for quantitative determination of trace levels of nitrosamines in gastric juice samples of patients with the gastrointestinal tract problems. The extraction conditions of each nitrosamine were optimized using regression modelling based on central composite design. The extraction conditions for all nitrosamines were selected to be 10.7 min for extraction time, 4.2 for pH and 23 for 2-propanol percentage in extraction solution. The obtained recoveries of nitrosamines ranged from 94.0 (NDMA) to 99.3 (NDPheA) %, and the precision of this method, as indicated by the relative standard deviations was within the range of 0.7 (NDPheA) and 2.6 (NDMA) %. The detection limits obtained from calculations by using GC-MS results based on S/N=3 were found within the range from 0.3 to 1.1 pg/mL. Total nitrosamine concentrations were found at the highest concentration up to 2431.12 pg/mL in cancer patients, whereas they were found at the lowest concentration down to 12.18 pg/mL in gastritis patients. The classification results of the gastric juice samples in different patient groups were very satisfactory, allowing 100% of patients to be correctly grouped. A new mathematical model has been developed allowing for the classification of gastric juices with a 93.1% success rate based on just the ratio of MNPIZ to DNPIZ. The ratio of MNPIZ to DNPIZ might be considered as a biomarker for the classification of gastric juices of patients and might act as an indicator of increased risk for stomach cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumsulfat, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumsulfat, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumphosphat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Natriumphosphat, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
Natriumsulfat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Natriumphosphat, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Natriumsulfat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
Sigma-Aldrich
Salzsäure -Lösung, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Natriumphosphat, meets USP testing specifications, anhydrous
Sigma-Aldrich
Natriumphosphat monobasisch -Lösung, BioUltra, 5 M in H2O
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%