Merck
  • Targeting FGFR Pathway in Human Hepatocellular Carcinoma: Expressing pFGFR and pMET for Antitumor Activity.
Targeting FGFR Pathway in Human Hepatocellular Carcinoma: Expressing pFGFR and pMET for Antitumor Activity.
Molecular cancer therapeutics (2015-09-10)
Jae-Cheol Jo, Eun Kyoung Choi, Jae-Sik Shin, Jai-Hee Moon, Seung-Woo Hong, Ha-Reum Lee, Seung-Mi Kim, Soo-A Jung, Dae-Hee Lee, Seang Hwan Jung, Sun-Hye Lee, Jeong Eun Kim, Kyu-pyo Kim, Yong Sang Hong, Young-Ah Suh, Se Jin Jang, Eun Kyung Choi, Jung Shin Lee, Dong-Hoon Jin, Tae Won Kim
ZUSAMMENFASSUNG
The MET receptor tyrosine kinase, the receptor for hepatocyte growth factor (HGF), has been implicated in cancer growth, invasion, migration, angiogenesis, and metastasis in a broad variety of human cancers, including human hepatocellular carcinoma (HCC). Recently, MET was suggested to be a potential target for the personalized treatment of HCC with an active HGF-MET signaling pathway. However, the mechanisms of resistance to MET inhibitors need to be elucidated to provide effective treatment. Here, we show that HCC cells exhibit different sensitivities to the MET inhibitor PHA665752, depending on the phosphorylation status of FGFR. Treatment of cells expressing both phospho-FGFR and phospho-MET with the inhibitor PHA665752 did not cause growth inhibition and cell death, whereas treatment with AZD4547, a pan-FGFR inhibitor, resulted in decreased colony formation and cleavage of caspase-3. Moreover, silencing of endogenous FGFR1 and FGFR2 by RNAi of HCC cells expressing phospho-FGFR, phospho-FGFR2, and phospho-MET overcame the resistance to PHA665752 treatment. Treatment of primary cancer cells from patients with HCC expressing both phospho-FGFR and phospho-MET with PHA665752 did not induce cell death, whereas AZD4547 treatment induced cell death through the cleavage of caspase-3. In addition, treatment of cells resistant to PHA665752 with AZD4547 abrogated the activation of downstream effectors of cell growth, proliferation, and survival. On the basis of these results, we conclude that the FGFR pathway is critical for HCC survival, and that targeting this pathway with AZD4547 may be beneficial for the treatment of patients with HCC-expressing phospho-FGFR and phospho-MET.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis