Merck
  • Determination of creatinine, uric and ascorbic acid in bovine milk and orange juice by hydrophilic interaction HPLC.
Determination of creatinine, uric and ascorbic acid in bovine milk and orange juice by hydrophilic interaction HPLC.
Food chemistry (2015-04-07)
Ruiting Zuo, Si Zhou, Yuegang Zuo, Yiwei Deng
ZUSAMMENFASSUNG
Creatinine (Cr), uric (UA) and ascorbic acid (AA) are common constituents in human fluids. Their abnormal concentrations in human fluids are associated with various diseases. Thus, apart from the endogenous formation in human body, it is also important to examine their sources from food products. In this study, a rapid and accurate HILIC method was developed for simultaneous determination of Cr, UA and AA in bovine milk and orange juice. Milk samples were pretreated by protein precipitation, centrifugation and filtration, followed by HPLC separation and quantification using a Waters Spherisorb S5NH2 column. The developed method has been successfully applied to determine the concentration of UA, AA and Cr in milk and fruit juice samples. The milk samples tested were found to contain UA and creatinine in the concentration range of 24.1-86.0 and 5.07-11.2 μg mL(-1), respectively. The orange juices contain AA over 212 μg mL(-1).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Natriumphosphat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Sigma-Aldrich
Natriumphosphat, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
Harnsäure, ≥99%, crystalline
Sigma-Aldrich
Natriumphosphat, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Kreatinin, anhydrous, ≥98%
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbinsäure, meets USP testing specifications
Sigma-Aldrich
Natriumphosphat, meets USP testing specifications, anhydrous
Sigma-Aldrich
Natriumphosphat monobasisch -Lösung, BioUltra, 5 M in H2O
Sigma-Aldrich
L-Ascorbinsäure, reagent grade
Sigma-Aldrich
Natriumphosphat, BioXtra, ≥99.0%
Sigma-Aldrich
Natriumphosphat, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Harnsäure, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbinsäure, FCC, FG
Sigma-Aldrich
L-Ascorbinsäure, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Natriumphosphat monobasisch-16O4, 99.9 atom % 16O