Merck
  • Elevated Expression of CD160 and 2B4 Defines a Cytolytic HIV-Specific CD8+ T-Cell Population in Elite Controllers.
Elevated Expression of CD160 and 2B4 Defines a Cytolytic HIV-Specific CD8+ T-Cell Population in Elite Controllers.
The Journal of infectious diseases (2015-04-18)
Carolina Pombo, E John Wherry, Emma Gostick, David A Price, Michael R Betts
ZUSAMMENFASSUNG
During chronic human immunodeficiency virus (HIV) infection, virus-specific CD8(+) T cells become functionally exhausted. Unlike most chronically infected individuals, elite controllers of HIV retain CD8(+) T-cell polyfunctionality and cytolytic capacity. It remains unclear whether elite controllers manifest T-cell exhaustion similar to subjects with chronic progression of HIV infection. Here we assessed coexpression of PD-1, Lag-3, CD160, and 2B4 as a measure of T-cell exhaustion in a cohort of elite controllers and in chronic progressors. We found that elite controllers have a high proportion of potentially exhausted (PD1(+)CD160(+)2B4(+)) HIV-specific CD8(+) T cells that is comparable to the proportion in chronic progressors. However, elite controllers also harbor a population of HIV-specific CD160(+)2B4(+) CD8(+) T cells that correlates with cytolytic capacity, as measured by perforin expression, a population not commonly present in chronic progressors. We therefore propose that coexpression of CD160 and 2B4 delineates a population of cytolytic CD8(+) T cells important for the control of HIV.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
SAFC
L-Glutamin
Sigma-Aldrich
Brefeldin A aus Penicillium brefeldianum, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Brefeldin A aus Penicillium brefeldianum, ≥99% (HPLC and TLC), BioXtra, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Brefeldin A aus Penicillium brefeldianum, Ready Made Solution, from Penicillium brefeldianum, 10 mg/mL in DMSO
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®