Merck
Anti-metastatic outcome of isoform-specific prolactin receptor targeting in breast cancer.
Cancer letters (2015-06-23)
Tomohiro Yonezawa, Kuan-Hui Ethan Chen, Mrinal K Ghosh, Lorena Rivera, Riva Dill, Lisa Ma, Pedro A Villa, Mitsumori Kawaminami, Ameae M Walker
ZUSAMMENFASSUNG
Controversy exists concerning the role of the long prolactin receptor (PRLR) in the progression of breast cancer. By targeting pre-mRNA splicing, we succeeded in knocking down only the long PRLR in vivo, leaving the short forms unaffected. Using two orthotopic and highly-metastatic models of breast cancer, one of which was syngeneic (mouse 4T1) to allow assessment of tumor-immune interactions and one of which was endocrinologically humanized (human BT-474) to activate human PRLRs, we examined the effect of long PRLR knockdown on disease progression. In both models, knockdown dramatically inhibited metastatic spread to the lungs and liver and resulted in increased central death in the primary tumor. In the syngeneic model, immune infiltrates in metastatic sites were changed from innate inflammatory cells to lymphocytes, with an increase in the incidence of tumor-specific cytotoxic T cells. Long PRLR knockdown in three-dimensional culture induced apoptosis of tumor-initiating/cancer stem cells (death of 95% of cells displaying stem cell markers in 15 days). We conclude that the long PRLR plays an important role in breast cancer metastasis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Progesteron, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesteron, ≥99%
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
β-Estradiol, meets USP testing specifications
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Progesteron, meets USP testing specifications
Sigma-Aldrich
Progesteron, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Prolaktin human, recombinant, expressed in E. coli, lyophilized powder, BioReagent, suitable for cell culture, >97% (SDS-PAGE)
Sigma-Aldrich
Formaldehyd-12C -Lösung, 20% in H2O, 99.9 atom % 12C