Merck
A novel safety assessment strategy applied to non-selective extracts.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2015-03-21)
Sander Koster, Winfried Leeman, Elwin Verheij, Ellen Dutman, Leo van Stee, Lene Munch Nielsen, Stefan Ronsmans, Hub Noteborn, Lisette Krul
ZUSAMMENFASSUNG
A main challenge in food safety research is to demonstrate that processing of foodstuffs does not lead to the formation of substances for which the safety upon consumption might be questioned. This is especially so since food is a complex matrix in which the analytical detection of substances, and consequent risk assessment thereof, is difficult to determine. Here, a pragmatic novel safety assessment strategy is applied to the production of non-selective extracts (NSEs), used for different purposes in food such as for colouring purposes, which are complex food mixtures prepared from reference juices. The Complex Mixture Safety Assessment Strategy (CoMSAS) is an exposure driven approach enabling to efficiently assess the safety of the NSE by focussing on newly formed substances or substances that may increase in exposure during the processing of the NSE. CoMSAS enables to distinguish toxicologically relevant from toxicologically less relevant substances, when related to their respective levels of exposure. This will reduce the amount of work needed for identification, characterisation and safety assessment of unknown substances detected at low concentration, without the need for toxicity testing using animal studies. In this paper, the CoMSAS approach has been applied for elderberry and pumpkin NSEs used for food colouring purposes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Essigsäureanhydrid, ReagentPlus®, ≥99%
Supelco
N-Methyl-N-trimethylsilyltrifluoracetamid, synthesis grade
Sigma-Aldrich
Trifluoressigsäureanhydrid, ReagentPlus®, ≥99%
Sigma-Aldrich
Essigsäureanhydrid, ACS reagent, ≥98.0%
Supelco
N-Methyl-N-trimethylsilyltrifluoracetamid, BioReagent, for silylations, LiChropur
Sigma-Aldrich
D-(−)-Ribose, ≥99% (GC)
Sigma-Aldrich
Essigsäureanhydrid, 99.5%
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
Dicyclohexylphthalat, 99%
Sigma-Aldrich
1-Heptanol, 98%
Sigma-Aldrich
Octanal, 99%
Sigma-Aldrich
D-(−)-Ribose, 98%
Supelco
N-Methyl-bis-trifluoracetamid, derivatization grade (GC derivatization), LiChropur, ≥97.0% (GC)
Sigma-Aldrich
Decanal, ≥98% (GC), liquid
Sigma-Aldrich
D-(−)-Ribose, ≥98%
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
D-(−)-Ribose, BioReagent, suitable for cell culture
Sigma-Aldrich
Decanal, natural, ≥97%, FG
Sigma-Aldrich
Heptylalkohol, ≥97%, FCC, FG
Sigma-Aldrich
Octanal, natural, ≥95%, FCC, FG
Sigma-Aldrich
Decanal, FCC, FG
Sigma-Aldrich
Octanal, ≥95%, FCC, FG
Sigma-Aldrich
1-Heptanol, ≥99.5% (GC)
Sigma-Aldrich
Essigsäureanhydrid, Arxada quality, ≥99.5% (GC)
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC