Merck
EFFECTS OF MITOMYCIN C ON HUMAN CHROMOSOMES.
The Journal of cell biology (1964-11-01)
M M COHEN, M W SHAW
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, powder, BioReagent, suitable for cell culture