Merck
  • A Simplified Method for the Aspiration of Bone Marrow from Patients Undergoing Hip and Knee Joint Replacement for Isolating Mesenchymal Stem Cells and In Vitro Chondrogenesis.
A Simplified Method for the Aspiration of Bone Marrow from Patients Undergoing Hip and Knee Joint Replacement for Isolating Mesenchymal Stem Cells and In Vitro Chondrogenesis.
Bone marrow research (2016-04-09)
Subhash C Juneja, Sowmya Viswanathan, Milan Ganguly, Christian Veillette
ZUSAMMENFASSUNG
The procedure for aspiration of bone marrow from the femur of patients undergoing total knee arthroplasty (TKA) or total hip arthroplasty (THA) may vary from an OR (operating room) to OR based on the surgeon's skill and may lead to varied extent of clotting of the marrow and this, in turn, presents difficulty in the isolation of mesenchymal stem cells (MSCs) from such clotted bone marrow. We present a simple detailed protocol for aspirating bone marrow from such patients, isolation, and characterization of MSCs from the aspirated bone marrow specimens and show that the bone marrow presented no clotting or exhibited minimal clotting. This represents an economical source and convenient source of MSCs from bone marrow for use in regenerative medicine. Also, we presented the detailed protocol and showed that the MSCs derived from such bone marrow specimens exhibited MSCs characteristics and generated micromass cartilages, the recipe for regenerative medicine for osteoarthritis. The protocols we presented can be used as standard operating procedures (SOPs) by researchers and clinicians.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Prolin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
Linolsäure, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Ölrot O, certified by the BSC
Sigma-Aldrich
Monoklonaler Anti-Kollagen-Antikörper, Typ I in Maus hergestellte Antikörper, clone COL-1, ascites fluid
Sigma-Aldrich
Anti-Maus-IgG-Antikörper des Esels, Cy3-Konjugat, Spezies-adsorbiert, Chemicon®, from donkey
Sigma-Aldrich
Human Mesenchymal Stem Cell Characterization Kit, The Human Mesenchymal Stem Cell Characterization Kit contains a panel of positive & negative selection markers for the characterization of the mesenchymal stem cell population in human samples.