Merck
  • Effects of redox cycling compounds on DT diaphorase activity in the liver of rainbow trout (Oncorhynchus mykiss).
Effects of redox cycling compounds on DT diaphorase activity in the liver of rainbow trout (Oncorhynchus mykiss).
Comparative hepatology (2005-05-06)
Joachim Sturve, Eiríkur Stephensen, Lars Förlin
ZUSAMMENFASSUNG
BACKGROUND: DT diaphorase (DTD; NAD(P)H:quinone oxidoreductase; EC 1.6.99.2) catalyses the two electron reduction of quinones, thus preventing redox cycling and consequently quinone dependent production of reactive oxygen species. In rat and mouse, a wide range of chemicals including polyaromatic hydrocarbons, azo dyes and quinones induces DTD. Bifunctional compounds, such as beta-naphthoflavone (beta-NF) and benzo(a)pyrene (B(a)P), induce DTD together with CYP1A and phase II enzymes by a mechanism involving the aryl hydrocarbon receptor (AHR). Monofunctional induction of DTD is mediated through the antioxidant response element and does not lead to the induction of AHR dependent enzymes, such as CYP1A. The aim of this study was to investigate the effects of prooxidants (both bifunctional and monofunctional) on the activity of hepatic DTD in rainbow trout (Oncorhynchus mykiss) in order to evaluate DTD suitability as a biomarker. We also investigated the effect of beta-NF on hepatic DTD activity in perch (Perca fluviatilis), shorthorn sculpin (Myoxocephalus scorpius), eelpout (Zoarces viviparus), brown trout (Salmo trutta) and carp (Cyprinus carpio). In addition, the effect of short term exposure to prooxidants on catalase activity was investigated. RESULTS: In rainbow trout, hepatic DTD activity is induced by the bifunctional AHR agonists beta-NF and B(a)P and the monofunctional inducers naphthazarin, menadione and paraquat. Although exposure to both B(a)P and beta-NF led to a strong 7-ethoxyresorufin-O-deethylase (EROD) induction, none of the monofunctional compounds affected the rainbow trout EROD activity. DTD was not induced by beta-NF in any of the other fish species. Much higher DTD activities were observed in rainbow trout compared to the other fish species. Catalase activity was less responsive to short term exposure to prooxidants compared to DTD. CONCLUSION: Since rainbow trout hepatic DTD activity is inducible by both monofunctional and bifunctional inducers, it is suggested that rainbow trout DTD may be regulated by the same mechanisms, as in mammals. The fact that DTD is inducible in rainbow trout suggests that the enzyme may be suitable as a part of a biomarker battery when rainbow trout is used in environmental studies. It appears as if DTD activity in rainbow trout is higher and inducible compared to the other fish species studied.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
5,8-Dihydroxy-1,4-naphthochinon, technical grade