Merck
  • A CEP215-HSET complex links centrosomes with spindle poles and drives centrosome clustering in cancer.
A CEP215-HSET complex links centrosomes with spindle poles and drives centrosome clustering in cancer.
Nature communications (2016-03-19)
Pavithra L Chavali, Gayathri Chandrasekaran, Alexis R Barr, Péter Tátrai, Chris Taylor, Evaggelia K Papachristou, C Geoffrey Woods, Sreenivas Chavali, Fanni Gergely
ZUSAMMENFASSUNG
Numerical centrosome aberrations underlie certain developmental abnormalities and may promote cancer. A cell maintains normal centrosome numbers by coupling centrosome duplication with segregation, which is achieved through sustained association of each centrosome with a mitotic spindle pole. Although the microcephaly- and primordial dwarfism-linked centrosomal protein CEP215 has been implicated in this process, the molecular mechanism responsible remains unclear. Here, using proteomic profiling, we identify the minus end-directed microtubule motor protein HSET as a direct binding partner of CEP215. Targeted deletion of the HSET-binding domain of CEP215 in vertebrate cells causes centrosome detachment and results in HSET depletion at centrosomes, a phenotype also observed in CEP215-deficient patient-derived cells. Moreover, in cancer cells with centrosome amplification, the CEP215-HSET complex promotes the clustering of extra centrosomes into pseudo-bipolar spindles, thereby ensuring viable cell division. Therefore, stabilization of the centrosome-spindle pole interface by the CEP215-HSET complex could promote survival of cancer cells containing supernumerary centrosomes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-α-Tubulin−-FITC-Antikörper, monoklonaler Antikörper der Maus, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Centrin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
MISSION® esiRNA, targeting human KIFC1