Merck
Activation profiles of opioid ligands in HEK cells expressing delta opioid receptors.
BMC neuroscience (2002-11-20)
Parham Gharagozlou, Hasan Demirci, J David Clark, Jelveh Lameh
ZUSAMMENFASSUNG
The aim of the present study was to characterize the activation profiles of 15 opioid ligands in transfected human embryonic kidney cells expressing only delta opioid receptors. Activation profiles of most of these ligands at delta opioid receptors had not been previously characterized in vitro. Receptor activation was assessed by measuring the inhibition of forskolin-stimulated cAMP production. Naltrexone and nalorphine were classified as antagonists at delta opioid receptor. The other ligands studied were agonists at delta opioid receptors and demonstrated IC50 values of 0.1 nM to 2 microM, maximal inhibition of 39-77% and receptor binding affinities of 0.5 to 243 nM. The rank order of efficacy of the ligands tested was metazocine = xorphanol > or = fentanyl = SKF 10047 = etorphine = hydromorphone = butorphanol = lofentanil > WIN 44,441 = Nalbuphine = cyclazocine > or = met-enkephalin > morphine = dezocine. For the first time these data describe and compare the function and relative efficacy of several ligands at delta opioid receptors. The data produced from this study can lead to elucidation of the complete activation profiles of several opioid ligands, leading to clarification of the mechanisms involved in physiological effects of these ligands at delta opioid receptors. Furthermore, these data can be used as a basis for novel use of existing opioid ligands based on their pharmacology at delta opioid receptors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
[D-Pen2,5]-Enkephalin hydrate, ≥95% (HPLC)
Sigma-Aldrich
(+)-N-Allylnormetazocine hydrochloride, ≥98% (HPLC)