- Aurora A phosphorylation of TACC3/maskin is required for centrosome-dependent microtubule assembly in mitosis.
Aurora A phosphorylation of TACC3/maskin is required for centrosome-dependent microtubule assembly in mitosis.
The Journal of cell biology (2005-09-21)
Kazuhisa Kinoshita, Tim L Noetzel, Laurence Pelletier, Karl Mechtler, David N Drechsel, Anne Schwager, Mike Lee, Jordan W Raff, Anthony A Hyman
PMID16172205
ZUSAMMENFASSUNG
Centrosomes act as sites of microtubule growth, but little is known about how the number and stability of microtubules emanating from a centrosome are controlled during the cell cycle. We studied the role of the TACC3-XMAP215 complex in this process by using purified proteins and Xenopus laevis egg extracts. We show that TACC3 forms a one-to-one complex with and enhances the microtubule-stabilizing activity of XMAP215 in vitro. TACC3 enhances the number of microtubules emanating from mitotic centrosomes, and its targeting to centrosomes is regulated by Aurora A-dependent phosphorylation. We propose that Aurora A regulation of TACC3 activity defines a centrosome-specific mechanism for regulation of microtubule polymerization in mitosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, clone DM1A, ascites fluid