Merck
  • The protein synthesis inhibitor blasticidin s enters mammalian cells via leucine-rich repeat-containing protein 8D.
The protein synthesis inhibitor blasticidin s enters mammalian cells via leucine-rich repeat-containing protein 8D.
The Journal of biological chemistry (2014-05-02)
Clarissa C Lee, Elizaveta Freinkman, David M Sabatini, Hidde L Ploegh
ZUSAMMENFASSUNG
Leucine-rich repeat-containing 8 (LRRC8) proteins have been identified as putative receptors involved in lymphocyte development and adipocyte differentiation. They remain poorly characterized, and no specific function has been assigned to them. There is no consensus on how this family of proteins might function because homology searches suggest that members of the LRRC8 family act not as plasma membrane receptors, but rather as channels that mediate cell-cell signaling. Here we provide experimental evidence that supports a role for LRRC8s in the transport of small molecules. We show that LRRC8D is a mammalian protein required for the import of the antibiotic blasticidin S. We characterize localization and topology of LRRC8A and LRRC8D and demonstrate that LRRC8D interacts with LRRC8A, LRRC8B, and LRRC8C. Given the suggested involvement in solute transport, our results support a model in which LRRC8s form one or more complexes that may mediate cell-cell communication by transporting small solutes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
ANTI-FLAG® M2-Affinitätsgel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoklonale ANTI-FLAG® M2-Peroxidase (HRP) in Maus hergestellte Antikörper, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Roche
Anti-HA-Peroxidase mit hoher Affinität, from rat IgG1
Roche
Anti-HA-Affinitätsmatrix, from rat IgG1