Merck
  • A benzenediamine derivate FC-99 attenuates lupus nephritis in MRL/lpr mice via inhibiting myeloid dendritic cell-secreted BAFF.
A benzenediamine derivate FC-99 attenuates lupus nephritis in MRL/lpr mice via inhibiting myeloid dendritic cell-secreted BAFF.
Acta biochimica et biophysica Sinica (2016-04-29)
Jianjian Ji, Jingjing Xu, Fanlin Li, Xiaojing Li, Wei Gong, Yuxian Song, Huan Dou, Yayi Hou
ZUSAMMENFASSUNG
Myeloid dendritic cells (DCs) can produce B-cell-activating factor (BAFF) that modulates survival and differentiation of B cells and plays a pivotal role in the pathogenesis of systemic lupus erythematosus (SLE). Toll-like receptor 4 (TLR4) signaling has important functions in the process of BAFF production. Our previous study showed that a benzenediamine derivate FC-99 possesses anti-inflammation activity and directly interacts with interleukin-1 receptor-associated kinase 4 (IRAK4), which was a pivotal molecule in TLR4 signaling. In this study, we demonstrated that FC-99 attenuated lupus nephritis in the MRL/lpr mice. FC-99 also decreased the levels of total immunoglobulin G (IgG), total IgG2a and IgM in sera, as well as the activation of B cells in the spleens of MRL/lpr mice. Moreover, FC-99 inhibited abnormal activation of myeloid DCs in spleens and reduced the levels of BAFF in sera, spleens, and kidneys of MRL/lpr mice. Furthermore, upon TLR4 stimulation with lipopolysaccharide in vitro, FC-99 inhibited IRAK4 phosphorylation, as well as the activation and BAFF production in murine bone marrow-derived DCs. These data indicate that FC-99 attenuates lupus nephritis in MRL/lpr mice via inhibiting DC-secreted BAFF, suggesting that FC-99 may be a potential therapeutic candidate for the treatment of SLE.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
FC-99 hydrochloride, ≥98% (HPLC)