Merck
  • AUTEN-67, an autophagy-enhancing drug candidate with potent antiaging and neuroprotective effects.
AUTEN-67, an autophagy-enhancing drug candidate with potent antiaging and neuroprotective effects.
Autophagy (2015-08-28)
Diána Papp, Tibor Kovács, Viktor Billes, Máté Varga, Anna Tarnóci, László Hackler, László G Puskás, Hanna Liliom, Krisztián Tárnok, Katalin Schlett, Adrienn Borsy, Zsolt Pádár, Attila L Kovács, Krisztina Hegedűs, Gábor Juhász, Marcell Komlós, Attila Erdős, Balázs Gulyás, Tibor Vellai
ZUSAMMENFASSUNG
Autophagy is a major molecular mechanism that eliminates cellular damage in eukaryotic organisms. Basal levels of autophagy are required for maintaining cellular homeostasis and functioning. Defects in the autophagic process are implicated in the development of various age-dependent pathologies including cancer and neurodegenerative diseases, as well as in accelerated aging. Genetic activation of autophagy has been shown to retard the accumulation of damaged cytoplasmic constituents, delay the incidence of age-dependent diseases, and extend life span in genetic models. This implies that autophagy serves as a therapeutic target in treating such pathologies. Although several autophagy-inducing chemical agents have been identified, the majority of them operate upstream of the core autophagic process, thereby exerting undesired side effects. Here, we screened a small-molecule library for specific inhibitors of MTMR14, a myotubularin-related phosphatase antagonizing the formation of autophagic membrane structures, and isolated AUTEN-67 (autophagy enhancer-67) that significantly increases autophagic flux in cell lines and in vivo models. AUTEN-67 promotes longevity and protects neurons from undergoing stress-induced cell death. It also restores nesting behavior in a murine model of Alzheimer disease, without apparent side effects. Thus, AUTEN-67 is a potent drug candidate for treating autophagy-related diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Freund-Adjuvans, komplett, cell suspension
Sigma-Aldrich
Freund-Adjuvans, inkomplett, liquid
Sigma-Aldrich
G 418 -disulfat (Salz) -Lösung, 50 mg/mL in H2O, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Bafilomycin A1 aus Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Cytosin β-D-Arabinofuranosid -hydrochlorid, crystalline
Sigma-Aldrich
Malachitgrün Oxalat, certified by the BSC
Sigma-Aldrich
Anti-GAPDH in Kaninchen hergestellte Antikörper, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Kaninchen-IgG (Gesamtmolekül)–Alkalische Phosphatase in Ziege hergestellte Antikörper, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
bis-Benzimid H 33258, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC)
Sigma-Aldrich
Anti-p62/SQSTM1 (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rat IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Maus-IgG (Gesamtmolekül)−Alkalische Phosphatase in Ziege hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution