Merck
  • Viable RNaseH1 knockout mice show RNaseH1 is essential for R loop processing, mitochondrial and liver function.
Viable RNaseH1 knockout mice show RNaseH1 is essential for R loop processing, mitochondrial and liver function.
Nucleic acids research (2016-05-01)
Walt F Lima, Heather M Murray, Sagar S Damle, Christopher E Hart, Gene Hung, Cheryl Li De Hoyos, Xue-Hai Liang, Stanley T Crooke
ZUSAMMENFASSUNG
Viable constitutive and tamoxifen inducible liver-specific RNase H1 knockout mice that expressed no RNase H1 activity in hepatocytes showed increased R-loop levels and reduced mitochondrial encoded DNA and mRNA levels, suggesting impaired mitochondrial R-loop processing, transcription and mitochondrial DNA replication. These changes resulted in mitochondrial dysfunction with marked changes in mitochondrial fusion, fission, morphology and transcriptional changes reflective of mitochondrial damage and stress. Liver degeneration ensued, as indicated by apoptosis, fibrosis and increased transaminase levels. Antisense oligonucleotides (ASOs) designed to serve as substrates for RNase H1 were inactive in the hepatocytes from the RNase H1 knockout mice and in vivo, demonstrating that RNase H1 is necessary for the activity of DNA-like ASOs. During liver regeneration, a clone of hepatocytes that expressed RNase H1 developed and partially restored mitochondrial and liver function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Chromatin-Immunpräzipitations-Kit EZ-Magna ChIP® A/G, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A/G beads. Control primers included.