Merck
  • Cortisol/cortisone ratio and matrix metalloproteinase-9 activity are associated with pediatric primary hypertension.
Cortisol/cortisone ratio and matrix metalloproteinase-9 activity are associated with pediatric primary hypertension.
Journal of hypertension (2016-08-05)
Alejandro Martinez-Aguayo, Carmen Campino, Rene Baudrand, Cristian A Carvajal, Hernán García, Marlene Aglony, Rodrigo Bancalari, Lorena García, Carolina Loureiro, Andrea Vecchiola, Alejandra Tapia-Castillo, Carolina Valdivia, Sebastian Sanhueza, Cristobal A Fuentes, Carlos F Lagos, Sandra Solari, Fidel Allende, Alexis M Kalergis, Carlos E Fardella
ZUSAMMENFASSUNG
To identify novel biomarkers associated with pediatric primary hypertension. We recruited 350 participants (4-16 years). Anthropometric parameters and aldosterone, plasma renin activity, cortisol, cortisone, Homeostasis Model Assessment Insulin Resistance (HOMA-IR), high-sensitivity C-reactive protein, adiponectin, IL-6, plasminogen activator inhibitor type 1 levels and matrix metalloproteinase-9 and matrix metalloproteinase-2 (MMP-9 and MMP-2) activities were measured. Genomic DNA was isolated. Patients with altered glucose metabolism, severe obesity [BMI-SD score (BMI-SDS) > 2.5], renovascular disease, primary aldosteronism and apparent mineralocorticoid excess syndrome were excluded. In selected participants (n = 320), SBP was positively correlated with BMI-SDS (r = 0.382, P < 0.001), HOMA-IR (r = 0.211, P < 0.001), MMP-9 activity (r = 0.215, P < 0.001) and the cortisol/cortisone ratio (r = 0.231, P < 0.001). DBP showed similar correlations with these variables. No correlation was observed with aldosterone or plasma renin activity. Participants were categorized as hypertensive (n = 59) or nonhypertensive (n = 261). In the univariate analysis, hypertensive patients had higher BMI-SDS (P < 0.001), HOMA-IR (P < 0.001), high-sensitivity C-reactive protein (P < 0.001), MMP-9 activity (P < 0.001), plasminogen activator inhibitor type 1 (P < 0.001) and cortisol/cortisone ratio (P < 0.001) than nonhypertensive patients. Multiple regression analysis showed that the variables that remained associated with hypertension were higher BMI-SDS [odds ratio (OR) = 3.74; 95% confidence interval (CI) = 1.84-7.58], a higher cortisol/cortisone ratio (OR = 3.92; 95% CI = 1.98-7.71) and increased MMP-9 activity (OR = 4.23; 95% CI = 2.15-8.32). We report that MMP-9 activity and the cortisol/cortisone ratio were higher in pediatric primary hypertensive patients, and these associations were independent of the effect of obesity. The potential role of these novel biomarkers in predicting hypertension risk and blood pressure regulation warrants further investigation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kortison, ≥95%