Merck
  • Treatment with Oxidized Phospholipids Directly Inhibits Nonalcoholic Steatohepatitis and Liver Fibrosis Without Affecting Steatosis.
Treatment with Oxidized Phospholipids Directly Inhibits Nonalcoholic Steatohepatitis and Liver Fibrosis Without Affecting Steatosis.
Digestive diseases and sciences (2016-04-15)
Itzhak Mendel, Niva Yacov, Anat Shoham, Eti Ishai, Eyal Breitbart
ZUSAMMENFASSUNG
Previous studies demonstrated that toll-like receptors 4 and 2 (TLR-4 and TLR-2), which are expressed on liver-resident Kupffer, hepatic stellate cells, and circulating monocytes, play a role in nonalcoholic fatty liver disease. Lecinoxoids are oxidized phospholipids that antagonize TLR-2- and TLR-4-mediated activation of innate immune cells and inhibit monocyte migration. In this study, we tested the effect of two functionally different lecinoxoids on the development of nonalcoholic steatohepatitis and liver fibrosis in a mouse model. Two-day-old C57BL/6 mice were injected with streptozotocin and fed a high-fat diet from Week 4 after birth. At Week 6 post-birth, lecinoxoids VB-201 or VB-703 were given orally, once daily, for 3 weeks. Telmisartan was administered orally, once daily, for 3 weeks, as positive control. At experiment conclusion, biochemical indices were evaluated. HE stain and quantitative PCR were used to determine the extent of steatosis and steatohepatitis, and Sirius red stain was used to assess liver fibrosis. Treatment with lecinoxoids did not alter the concentration of blood glucose, liver triglycerides, or steatosis compared with solvent-treated mice. However, whereas VB-201 inhibited the development of fibrosis and, to some extent, liver inflammation, VB-703 significantly lessened both liver inflammation and fibrosis. This study indicates that using lecinoxoids to antagonize TLR-2, and more prominently TLR-4, is sufficient to significantly inhibit nonalcoholic steatohepatitis and liver fibrosis. Inhibiting monocyte migration with lecinoxoids that are relatively weak TLR-4 antagonists may alter liver fibrosis and to some extent nonalcoholic steatohepatitis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, clone DM1A, ascites fluid
Sigma-Aldrich
Monoklonale Anti-MAP-Kinase, aktiviert (Diphosphorylierter ERK-1&2) in Maus hergestellte Antikörper, clone MAPK-YT, ascites fluid