Merck
  • Novel protein kinase C isoforms regulate human keratinocyte differentiation by activating a p38 delta mitogen-activated protein kinase cascade that targets CCAAT/enhancer-binding protein alpha.
Novel protein kinase C isoforms regulate human keratinocyte differentiation by activating a p38 delta mitogen-activated protein kinase cascade that targets CCAAT/enhancer-binding protein alpha.
The Journal of biological chemistry (2002-06-25)
Tatiana Efimova, Anne Deucher, Toshio Kuroki, Motoi Ohba, Richard L Eckert
ZUSAMMENFASSUNG
The novel protein kinase C (nPKC) isoforms are important regulators of human involucrin (hINV) gene expression during keratinocyte differentiation (Efimova, T., and Eckert, R. L. (2000) J. Biol. Chem. 275, 1601-1607). Although the regulatory mechanism involves mitogen-activated protein kinase (MAPK) activation, the role of individual MAPK isoforms has not been elucidated. We therefore examined the effects of individual nPKCs on MAPK activation. We observe unique changes whereby nPKC expression simultaneously increases p38 activity and decreases ERK1 and ERK2 activity. Although p38 alpha, p38 beta, and p38 delta are expressed in keratinocytes, only a single isoform, p38 delta, accounts for the increased p38 activity. Parallel studies indicate that this isoform is also activated by treatment with the keratinocyte regulatory agents, 12-O-tetradecanoylphorbol-13-acetate, calcium, and okadaic acid. These changes in MAPK activity are associated with increased C/EBP alpha transcription factor expression and DNA binding to the hINV promoter and increased hINV gene expression. Expression of PKC delta, PKC epsilon, or PKC eta causes a 10-fold increase in hINV promoter activity, whereas C/EBP alpha expression produces a 25-fold increase. However, simultaneous expression of both proteins causes a synergistic 100-fold increase in promoter activity. These responses are eliminated by the dominant-negative C/EBP isoform, GADD153, and are also inhibited by dominant-negative forms of Ras, MEKK1, MEK3, and p38. These results suggest that the nPKC isoforms produce a unique shift in MAPK activity via a Ras, MEKK1, MEK3 pathway, to increase p38 delta and inhibit ERK1/2 and ultimately increase C/EBP alpha binding to the hINV promoter and hINV gene expression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoklonale ANTI-FLAG® M2-Peroxidase (HRP) in Maus hergestellte Antikörper, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-MAP Kinase (ERK-1, ERK-2) antibody produced in rabbit, whole antiserum