Merck
  • In vitro cytotoxic and genotoxic evaluation of peptides used in nuclear medicine (DOTATATE and Ubiquicidin
In vitro cytotoxic and genotoxic evaluation of peptides used in nuclear medicine (DOTATATE and Ubiquicidin
Cytotechnology (2016-10-01)
Ivette Zegarra Ocampo, Priscila de Queiroz Souza Passos, Luma Ramirez de Carvalho, Camila Ayala Lira da Cruz, Natália Mencacci Esteves-Pedro, Fabiana Medeiros da Silva, Olga Zazuco Higa, Luiz Alberto Pereira Dias, Kayo Okazaki, Daniel Perez Vieira
ZUSAMMENFASSUNG
Micronucleus (MN) assay constitutes a valuable surrogate to the chromosome aberration technique for in vitro testing of the genotoxicity of substances. As test substances, two peptidic compounds (DOTATATE and Ubiquicidin
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Cytochalasin B aus Drechslera dematioidea, ≥98% (HPLC), powder
Sigma-Aldrich
Colchicin, ≥95% (HPLC), powder
Sigma-Aldrich
Acridinorange -hydrochlorid -Lösung, 10 mg/mL in H2O, ≥95.0% (HPLC)