Merck
  • A simplified approach for efficient isolation of functional microglial cells: Application for modeling neuroinflammatory responses in vitro.
A simplified approach for efficient isolation of functional microglial cells: Application for modeling neuroinflammatory responses in vitro.
Glia (2016-07-28)
Julia E Sepulveda-Diaz, Mohand O Ouidja, Sergio B Socias, Sabah Hamadat, Serge Guerreiro, Rita Raisman-Vozari, Patrick P Michel
ZUSAMMENFASSUNG
Purified microglial cells in culture are frequently used to model brain inflammatory responses but obtaining large yields of these cells on a routine basis can be quite challenging. Here, we demonstrate that it is possible to achieve high-yield isolation of pure microglial (MAC-1(+) /Fcrls(+) /Ccr2(-) ) cells from postnatal brain tissue through a simple culture procedure that mainly relies on the adhesion preference of these cells to the polycation polyethyleneimine (PEI) in serum-supplemented DMEM medium. Accordingly, other synthetic or biological substrates failed to mimic PEI effects under the same culture conditions. Replacement of DMEM by DMEM/F12 nutrient mixture did not permit microglial cell isolation on PEI coating, indicating that PEI effects were context-dependent. Remarkably, the lack of culture feeding during progression of microglial cell isolation strongly improved cell yield, suggesting that nutritional deprivation was required to optimize this process. When generated in large culture flasks coated with PEI, cultures of microglial cells were easily recovered by trypsin proteolysis to produce subcultures for functional studies. These cultures responded to lipopolysaccharide (LPS, 1-10 ng/ml) treatment by secreting pro-inflammatory cytokines such as TNF-α, IL-6, IL-1β and by generating nitric oxide and reactive oxygen species. Most interestingly, this response was curtailed by appropriate reference drugs. Microglial cells were also strongly responsive to the mitogenic cytokine GM-CSF, which confirms that the functional repertoire of these cells was well preserved. Because of its high yield and simplicity, we believe that the present method will prove to be especially convenient for mechanistic studies or screening assays. GLIA 2016;64:1912-1924.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly-L-ornithin -Lösung, mol wt 30,000-70,000, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Supelco
Polyethylenimin -Lösung, analytical standard, 50 % (w/v) in H2O
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O26:B6, ≥10,000 EU/mg, purified by phenol extraction