Merck
Autophagy modulates SNCA/α-synuclein release, thereby generating a hostile microenvironment.
Autophagy (2014-12-09)
Anne-Maria Poehler, Wei Xiang, Philipp Spitzer, Verena Elisabeth Luise May, Holger Meixner, Edward Rockenstein, Oldriska Chutna, Tiago Fleming Outeiro, Juergen Winkler, Eliezer Masliah, Jochen Klucken
ZUSAMMENFASSUNG
SNCA/α-synuclein aggregation plays a crucial role in synucleinopathies such as Parkinson disease and dementia with Lewy bodies. Aggregating and nonaggregating SNCA species are degraded by the autophagy-lysosomal pathway (ALP). Previously, we have shown that the ALP is not only responsible for SNCA degradation but is also involved in the intracellular aggregation process of SNCA. An additional role of extracellular SNCA in the pathology of synucleinopathies substantiating a prion-like propagation hypothesis has been suggested since released SNCA species and spreading of SNCA pathology throughout neural cells have been observed. However, the molecular interplay between intracellular pathways, SNCA aggregation, release, and response of the local microenvironment remains unknown. Here, we attributed SNCA-induced toxicity mainly to secreted species in a cell culture model of SNCA aggregation and in SNCA transgenic mice: We showed that ALP inhibition by bafilomycinA1 reduced intracellular SNCA aggregation but increased secretion of smaller oligomers that exacerbated microenvironmental response including uptake, inflammation, and cellular damage. Low-aggregated SNCA was predominantly released by exosomes and RAB11A-associated pathways whereas high-aggregated SNCA was secreted by membrane shedding. In summary, our study revealed a novel role of the ALP by linking protein degradation to nonclassical secretion for toxic SNCA species. Thus, impaired ALP in the diseased brain not only limits intracellular degradation of misfolded proteins, but also leads to a detrimental microenvironmental response due to enhanced SNCA secretion. These findings suggest that the major toxic role of SNCA is related to its extracellular species and further supports a protective role of intracellular SNCA aggregation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
cOmplete, Mini, EDTA-freier Protease-Inhibitor-Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Trizma® Base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Deoxycholsäure Natriumsalz, ≥97% (titration)
Sigma-Aldrich
Anti-NeuN-Antikörper, Klon A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Chloroquin -diphosphat (Salz), powder or crystals, 98.5-101.0% (EP), meets EP testing specifications
Sigma-Aldrich
Paraformaldehyd, prilled, 95%
Sigma-Aldrich
Anti-Gliafaserprotein-Antikörper, Klon GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
3-Methyladenin, autophagy inhibitor
Sigma-Aldrich
Anti-Synuclein-α-Antikörper, serum, Chemicon®
Sigma-Aldrich
Anti-α-Synuclein-Antikörper, Klon Syn211, ascites fluid, clone Syn211, Upstate®