Merck
  • Neutrophil elastase-initiated EGFR/MEK/ERK signaling counteracts stabilizing effect of autocrine TGF-beta on tropoelastin mRNA in lung fibroblasts.
Neutrophil elastase-initiated EGFR/MEK/ERK signaling counteracts stabilizing effect of autocrine TGF-beta on tropoelastin mRNA in lung fibroblasts.
American journal of physiology. Lung cellular and molecular physiology (2006-02-14)
Sandra J DiCamillo, Shenghong Yang, Maria V Panchenko, Paul A Toselli, Estee F Naggar, Celeste B Rich, Phillip J Stone, Matthew A Nugent, Mikhail P Panchenko
ZUSAMMENFASSUNG
Neutrophil elastase (NE) plays an important role in emphysema, a pulmonary disease associated with excessive elastolysis and ineffective repair of interstitial elastin. Besides its direct elastolytic activity, NE releases soluble epidermal growth factor receptor (EGFR) ligands and initiates EGFR/MEK/ERK signaling to downregulate tropoelastin mRNA in neonatal rat lung fibroblasts (DiCamillo SJ, Carreras I, Panchenko MV, Stone PJ, Nugent MA, Foster JA, and Panchenko MP. J Biol Chem 277: 18938-18946, 2002). We now report that NE downregulates tropoelastin mRNA in the rat fetal lung fibroblast line RFL-6. The tropoelastin mRNA downregulation is preceded by release of EGF-like and TGF-alpha-like polypeptides and requires EGFR/MEK/ERK signaling, because it is prevented by the EGFR inhibitor AG1478 and the MEK/ERK uncoupler U0126. Tropoelastin expression in RFL-6 fibroblasts is governed by autocrine TGF-beta signaling, because TGF-beta type I receptor kinase inhibitor or TGF-beta neutralizing antibody dramatically decreases tropoelastin mRNA and protein levels. Half-life of tropoelastin mRNA in RFL-6 cells is >24 h, but it is decreased to approximately 8 h by addition of TGF-beta neutralizing antibody, EGF, TGF-alpha, or NE. Tropoelastin mRNA destabilization by NE, EGF, or TGF-alpha is abolished by AG1478 or U0126. EGF-dependent tropoelastin mRNA downregulation is reversed upon ligand withdrawal, whereas chronic EGF treatment leads to persistent downregulation of tropoelastin mRNA and protein levels and decreases insoluble elastin deposition. We conclude that NE-initiated EGFR/MEK/ERK signaling cascade overrides the autocrine TGF-beta signaling on tropoelastin mRNA stability and, therefore, decreases the elastogenic response in RFL-6 fibroblasts. We hypothesize that persistent EGFR/MEK/ERK signaling could impede the TGF-beta-induced elastogenesis/elastin repair in the chronically inflamed, elastase/anti-elastase imbalanced lung in emphysema.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Kaninchen-IgG-(Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Smad2/3 Antibody, Upstate®, from rabbit