Merck
Liquid crystal-based biosensor with backscattering interferometry: A quantitative approach.
Biosensors & bioelectronics (2016-09-27)
Mashooq Khan, Soo-Young Park
ZUSAMMENFASSUNG
We developed a new technology that uses backscattering interferometry (BSI) to quantitatively measure nematic liquid crystal (NLC)-based biosensors, those usually relied on texture reading for on/off signals. The LC-based BSI comprised an octadecyltrichlorosilane (OTS)-coated square capillary filled with 4-cyano-4'-pentylbiphenyl (5CB, a nematic LC at room temperature). The LC/water interface in the capillary was functionalized by a coating of poly(acrylicacid-b-4-cyanobiphenyl-4'-oxyundecylacrylate) (PAA-b-LCP) and immobilized with the enzymes glucose oxidase (GOx) and horseradish peroxidase (HRP) through covalent linkage to the PAA chains (5CBPAA-GOx:HRP) for glucose detection. Laser irradiation of the LC near the LC/water interface resulted in backscattered fringes with high contrast. The change in the spatial position of the fringes (because of the change in the orientation of the LC caused by the GOx:HRP enzymatic reaction of glucose) altered the output voltage of the photodetector when its active area was aligned with the edge of one of the fringes. The change in the intensity at the photodetector allowed the detection limit of the instrument to be as low as 0.008mM with a linear range of 0.02-9mM in a short response time (~60s). This LC-based BSI technique allows for quantitative, sensitive, selective, reproducible, easily obtainable, and interference-free detection in a large linear dynamic range and for practical applications with human serum.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Hydroxysulfosuccinimid Natriumsalz, ≥98% (HPLC)
Sigma-Aldrich
Trichlor(octadecyl)silan, ≥90%