Merck
  • Bayesian quantile regression-based nonlinear mixed-effects joint models for time-to-event and longitudinal data with multiple features.
Bayesian quantile regression-based nonlinear mixed-effects joint models for time-to-event and longitudinal data with multiple features.
Statistics in medicine (2016-09-07)
Yangxin Huang, Jiaqing Chen
ZUSAMMENFASSUNG
This article explores Bayesian joint models for a quantile of longitudinal response, mismeasured covariate and event time outcome with an attempt to (i) characterize the entire conditional distribution of the response variable based on quantile regression that may be more robust to outliers and misspecification of error distribution; (ii) tailor accuracy from measurement error, evaluate non-ignorable missing observations, and adjust departures from normality in covariate; and (iii) overcome shortages of confidence in specifying a time-to-event model. When statistical inference is carried out for a longitudinal data set with non-central location, non-linearity, non-normality, measurement error, and missing values as well as event time with being interval censored, it is important to account for the simultaneous treatment of these data features in order to obtain more reliable and robust inferential results. Toward this end, we develop Bayesian joint modeling approach to simultaneously estimating all parameters in the three models: quantile regression-based nonlinear mixed-effects model for response using asymmetric Laplace distribution, linear mixed-effects model with skew-t distribution for mismeasured covariate in the presence of informative missingness and accelerated failure time model with unspecified nonparametric distribution for event time. We apply the proposed modeling approach to analyzing an AIDS clinical data set and conduct simulation studies to assess the performance of the proposed joint models and method. Copyright © 2016 John Wiley & Sons, Ltd.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)