Merck
  • Rapid endoglin determination in serum samples using an amperometric magneto-actuated disposable immunosensing platform.
Rapid endoglin determination in serum samples using an amperometric magneto-actuated disposable immunosensing platform.
Journal of pharmaceutical and biomedical analysis (2016-07-23)
Rebeca M Torrente-Rodríguez, Susana Campuzano, Víctor Ruiz-Valdepeñas-Montiel, María Pedrero, M Jesús Fernández-Aceñero, Rodrigo Barderas, José M Pingarrón
ZUSAMMENFASSUNG
A sensitive and rapid method for the determination of the clinically relevant biomarker human endoglin (CD105) in serum samples is presented, involving a magneto-actuated immunoassay and amperometric detection at disposable screen-printed carbon electrodes (SPCEs). Micro-sized magnetic particles were modified with a specific antibody to selectively capture the target protein which was further sandwiched with a secondary HRP-labeled antibody. The immunocomplexes attached to the magnetic carriers were amperometrically detected at SPCEs using the hydroquinone (HQ)/H2O2/HRP system. The magneto-actuated immunosensing platform was able to detect 5 pmoles of endoglin (in 25μL of sample, 0.2μM) in 30min providing statistically similar results to those obtained using a commercial ELISA kit for the determination of endogenous content of endoglin in human serum samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Hydroxysulfosuccinimid Natriumsalz, ≥98% (HPLC)
Sigma-Aldrich
Hämoglobin human, lyophilized powder