Merck
Prognostic Utility of a New mRNA Expression Signature of Gleason Score.
Clinical cancer research : an official journal of the American Association for Cancer Research (2016-09-25)
Jennifer A Sinnott, Sam F Peisch, Svitlana Tyekucheva, Travis Gerke, Rosina Lis, Jennifer R Rider, Michelangelo Fiorentino, Meir J Stampfer, Lorelei A Mucci, Massimo Loda, Kathryn L Penney
ZUSAMMENFASSUNG
Gleason score strongly predicts prostate cancer mortality; however, scoring varies among pathologists, and many men are diagnosed with intermediate-risk Gleason score 7. We previously developed a 157-gene signature for Gleason score using a limited gene panel. Using a new whole-transcriptome expression dataset, we verified the previous signature's performance and developed a new Gleason signature to improve lethal outcome prediction among men with Gleason score 7. We generated mRNA expression data from prostate tumor tissue from men in the Physicians' Health Study and Health Professionals Follow-Up Study (N = 404) using the Affymetrix Human Gene 1.0 ST microarray. The Prediction Analysis for Microarrays method was used to develop a signature to distinguish high (≥8) versus low (≤6) Gleason score. We evaluated the signature's ability to improve prediction of lethality among men with Gleason score 7, adjusting for 3 + 4/4 + 3 status, by quantifying the area under the receiver operating characteristic (ROC) curve (AUC). We identified a 30-gene signature that best distinguished Gleason score ≤6 from ≥8. The AUC to predict lethal disease among Gleason score 7 men was 0.76 [95% confidence interval (CI), 0.67-0.84] compared with 0.68 (95% CI, 0.59-0.76) using 3 + 4/4 + 3 status alone (P = 0.0001). This signature was a nonsignificant (P = 0.09) improvement over our previous signature (AUC = 0.72). Our new 30-gene signature improved prediction of lethality among men with Gleason score 7. This signature can potentially become a useful prognostic tool for physicians to improve treatment decision making. Clin Cancer Res; 23(1); 81-87. ©2016 AACRSee related commentary by Yin et al., p. 6.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phytosphingosine hydrochloride