Merck
  • The conserved protein Seb1 drives transcription termination by binding RNA polymerase II and nascent RNA.
The conserved protein Seb1 drives transcription termination by binding RNA polymerase II and nascent RNA.
Nature communications (2017-04-04)
Sina Wittmann, Max Renner, Beth R Watts, Oliver Adams, Miles Huseyin, Carlo Baejen, Kamel El Omari, Cornelia Kilchert, Dong-Hyuk Heo, Tea Kecman, Patrick Cramer, Jonathan M Grimes, Lidia Vasiljeva
ZUSAMMENFASSUNG
Termination of RNA polymerase II (Pol II) transcription is an important step in the transcription cycle, which involves the dislodgement of polymerase from DNA, leading to release of a functional transcript. Recent studies have identified the key players required for this process and showed that a common feature of these proteins is a conserved domain that interacts with the phosphorylated C-terminus of Pol II (CTD-interacting domain, CID). However, the mechanism by which transcription termination is achieved is not understood. Using genome-wide methods, here we show that the fission yeast CID-protein Seb1 is essential for termination of protein-coding and non-coding genes through interaction with S2-phosphorylated Pol II and nascent RNA. Furthermore, we present the crystal structures of the Seb1 CTD- and RNA-binding modules. Unexpectedly, the latter reveals an intertwined two-domain arrangement of a canonical RRM and second domain. These results provide important insights into the mechanism underlying eukaryotic transcription termination.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonale ANTI-FLAG® M2-Peroxidase (HRP) in Maus hergestellte Antikörper, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Kaninchen-IgG-(Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody
Sigma-Aldrich
Anti-Maus-IgG (Fab-spezifisch)–Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Peroxidase Anti-Peroxidase Soluble Complex antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-RNA-Polymerase II-Untereinheit B1(Phospho-CTD-Ser-5)-Antikörper, Klon 3E8, culture supernatant, clone 3E8, from rat
Sigma-Aldrich
Goat Anti-Rat IgG Peroxidase Conjugate, lyophilized, Calbiochem®
Sigma-Aldrich
Anti-RNA-Polymerase II-Untereinheit B1 (Phospho-CTD Ser-2), Klon 3E10 (Ratte, monoklonal)., culture supernatant, clone 3E10, from rat
Sigma-Aldrich
Anti-RNA-Polymerase II-Untereinheit B1 (Phospho-CTD Ser-7)-Antikörper, Klon 4E12, culture supernatant, clone 4E12, from rat