Merck
Mutants of Ca
Photochemistry and photobiology (2017-01-08)
Vasilisa V Krasitskaya, Ludmila P Burakova, Anastasia A Komarova, Eugenia E Bashmakova, Ludmila A Frank
ZUSAMMENFASSUNG
Color variants of Ca
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-(N-Maleimidomethyl)cyclohexan-Carboxylsäure N-hydroxysuccinimid-Ester, ≥98%, powder