- Interstrand DNA covalent binding of two dinuclear Ru(ii) complexes. Influence of the extra ring of the bridging ligand on the DNA interaction and cytotoxic activity.
Interstrand DNA covalent binding of two dinuclear Ru(ii) complexes. Influence of the extra ring of the bridging ligand on the DNA interaction and cytotoxic activity.
Dalton transactions (Cambridge, England : 2003) (2017-03-01)
Héctor J Lozano, Natalia Busto, Gustavo Espino, Arancha Carbayo, José M Leal, James A Platts, Begoña García
PMID28243638
ZUSAMMENFASSUNG
In this work, we report experimental and computational evidence for the intercalation into the DNA base-pairs of the free quinones quinizarin (Q) and naphthazarin (N) and the interstrand covalent binding of their p-cymene di-ruthenium(ii) complexes (Cl