- Inflammatory cytokines compromise programmed cell death-1 (PD-1)-mediated T cell suppression in inflammatory arthritis through up-regulation of soluble PD-1.
Inflammatory cytokines compromise programmed cell death-1 (PD-1)-mediated T cell suppression in inflammatory arthritis through up-regulation of soluble PD-1.
Clinical and experimental immunology (2017-03-01)
D Bommarito, C Hall, L S Taams, V M Corrigall
PMID28245522
ZUSAMMENFASSUNG
The programmed cell death 1 (PD-1) receptor plays a major role in regulating T cell activation. Our aim was to determine how inflammation influences PD-1-mediated T cell suppression. Flow cytometry analysis of rheumatoid arthritis (RA) and psoriatic arthritis (PsA) synovial fluid (SF) mononuclear cells showed an increase in the percentage of PD-1