Merck
  • All muscarinic acetylcholine receptors (M1-M5) are expressed in murine brain microvascular endothelium.
All muscarinic acetylcholine receptors (M1-M5) are expressed in murine brain microvascular endothelium.
Scientific reports (2017-07-13)
Beatrice Mihaela Radu, Antonio Marco Maria Osculati, Eda Suku, Adela Banciu, Grygoriy Tsenov, Flavia Merigo, Marzia Di Chio, Daniel Dumitru Banciu, Cristina Tognoli, Petr Kacer, Alejandro Giorgetti, Mihai Radu, Giuseppe Bertini, Paolo Francesco Fabene
ZUSAMMENFASSUNG
Clinical and experimental studies indicate that muscarinic acetylcholine receptors are potential pharmacological targets for the treatment of neurological diseases. Although these receptors have been described in human, bovine and rat cerebral microvascular tissue, a subtype functional characterization in mouse brain endothelium is lacking. Here, we show that all muscarinic acetylcholine receptors (M1-M5) are expressed in mouse brain microvascular endothelial cells. The mRNA expression of M2, M3, and M5 correlates with their respective protein abundance, but a mismatch exists for M1 and M4 mRNA versus protein levels. Acetylcholine activates calcium transients in brain endothelium via muscarinic, but not nicotinic, receptors. Moreover, although M1 and M3 are the most abundant receptors, only a small fraction of M1 is present in the plasma membrane and functions in ACh-induced Ca2+ signaling. Bioinformatic analyses performed on eukaryotic muscarinic receptors demonstrate a high degree of conservation of the orthosteric binding site and a great variability of the allosteric site. In line with previous studies, this result indicates muscarinic acetylcholine receptors as potential pharmacological targets in future translational studies. We argue that research on drug development should especially focus on the allosteric binding sites of the M1 and M3 receptors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
GenElute Säugetier-Gesamt-RNA-Miniprep-Kit, sufficient for 70 purifications
Sigma-Aldrich
Acetylcholinbromid, ≥99%
Sigma-Aldrich
4-DAMP, ≥98% (HPLC)
Sigma-Aldrich
Telenzepine dihydrochloride hydrate, solid, ≥98% (HPLC)
Sigma-Aldrich
AC 42, ≥98% (HPLC)