Merck
  • Cell culture media supplemented with raffinose reproducibly enhances high mannose glycan formation.
Cell culture media supplemented with raffinose reproducibly enhances high mannose glycan formation.
Journal of biotechnology (2017-05-04)
David Brühlmann, Anais Muhr, Rebecca Parker, Thomas Vuillemin, Blanka Bucsella, Franka Kalman, Serena Torre, Fabio La Neve, Antonio Lembo, Tobias Haas, Markus Sauer, Jonathan Souquet, Hervé Broly, Jürgen Hemberger, Martin Jordan
ZUSAMMENFASSUNG
Glycosylation plays a pivotal role in pharmacokinetics and protein physiochemical characteristics. In particular, effector functions including antibody-dependent cell-mediated cytotoxicity (ADCC) can be desired, and it has been described that high-mannose species exhibited enhanced ADCC. In this work we present the trisaccharide raffinose as a novel cell culture medium supplement to promote high mannose N-glycans in fed-batch cultures, which is sought after in the development of biosimilars to match the quality profile of the reference medicinal product (RMP) also. Up to six-fold increases of high mannose species were observed with increasing raffinose concentrations in the medium of shaken 96-deepwell plates and shake tubes when culturing two different CHO cell lines in two different media. The findings were confirmed in a pH-, oxygen- and CO
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-(−)-Glukose, ≥99%
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)