Merck
  • Prostaglandin E receptor subtype 4 regulates lipid droplet size and mitochondrial activity in murine subcutaneous white adipose tissue.
Prostaglandin E receptor subtype 4 regulates lipid droplet size and mitochondrial activity in murine subcutaneous white adipose tissue.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2017-05-24)
Fan Ying, Yin Cai, Yu Cai, Yu Wang, Eva Hoi Ching Tang
ZUSAMMENFASSUNG
The purpose of this study was to investigate whether genetic ablation of prostaglandin E receptor subtype 4 (EP4) affects white adipose tissue (WAT) remodeling mediated by β
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)
Sigma-Aldrich
Coenzym Q1, ≥95%