Merck
  • JAK inhibitor has the amelioration effect in lupus-prone mice: the involvement of IFN signature gene downregulation.
JAK inhibitor has the amelioration effect in lupus-prone mice: the involvement of IFN signature gene downregulation.
BMC immunology (2017-08-24)
Keigo Ikeda, Kunihiro Hayakawa, Maki Fujishiro, Mikiko Kawasaki, Takuya Hirai, Hiroshi Tsushima, Tomoko Miyashita, Satoshi Suzuki, Shinji Morimoto, Naoto Tamura, Kenji Takamori, Hideoki Ogawa, Iwao Sekigawa
ZUSAMMENFASSUNG
We previously reported that JAK-STAT-pathway mediated regulation of IFN-regulatory factor genes could play an important role in SLE pathogenesis. Here, we evaluated the efficacy of the JAK inhibitor tofacitinib (TOFA) for controlling IFN signalling via the JAK-STAT pathway and as a therapeutic for SLE. We treated NZB/NZW F1 mice with TOFA and assessed alterations in their disease, pathological, and immunological conditions. Gene-expression results obtained from CD4 Modulation of type I IFN signalling via JAK-STAT inhibition may exert a beneficial effect in SLE patients, and our results suggest that TOFA could be utilised for the development of new SLE-specific therapeutic strategies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyrogallol, Rot, Suitable for use as a complexometric indicator
Sigma-Aldrich
TOFA, ≥98% (HPLC)