Merck
Regulation of retinoic acid synthetic enzymes by WT1 and HDAC inhibitors in 293 cells.
International journal of molecular medicine (2017-07-06)
Yifan Li, Lei Wang, Weipeng Ai, Nianhui He, Lin Zhang, Jihui Du, Yong Wang, Xingjian Mao, Junqi Ren, Dan Xu, Bei Zhou, Rong Li, Liwen Mai
ZUSAMMENFASSUNG
All-trans retinoic acid (atRA), which is mainly generated endogenously via two steps of oxidation from vitamin A (retinol), plays an indispensible role in the development of the kidney and many other organs. Enzymes that catalyze the oxidation of retinol to generate atRA, including aldehyde dehydrogenase 1 family (ALDH1)A1, ALDH1A2 and ALDH1A3, exhibit complex expression patterns at different stages of renal development. However, molecular triggers that control these differential expression levels are poorly understood. In this study, we provide in vitro evidence to demonstrate that Wilms' tumor 1 (WT1) negatively regulates the expression of the atRA synthetic enzymes, ALDH1A1, ALDH1A2 and ALDH1A3, in the 293 cell line, leading to significant blockage of atRA production. Furthermore, we demonstrate that the suppression of ALDH1A1 by WT1 can be markedly attenuated by histone deacetylase inhibitors (HDACis). Taken together, we provide evidence to indicate that WT1 and HDACs are strong regulators of endogenous retinoic acid synthetic enzymes in 293 cells, indicating that they may be involved in the regulation of atRA synthesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Retinsäure, ≥98% (HPLC), powder
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
5-Aza-2′-Desoxycytidin, ≥97%
Sigma-Aldrich
SAHA, ≥98% (HPLC)
Sigma-Aldrich
DL-Glyceraldehyd-3-phosphat -Lösung, 45-55 mg/mL in H2O