Merck
  • Lymphoproliferative responses to dendritic cell presentation of sensitizing allergens in atopic children with multiple allergies.
Lymphoproliferative responses to dendritic cell presentation of sensitizing allergens in atopic children with multiple allergies.
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology (2017-09-12)
Tim H Scott-Taylor, Stefan-Claudiu Axinia, Stephan Strobel
ZUSAMMENFASSUNG
Peripheral blood mononuclear cells (PBMCs) proliferate inconsistently, rendering current lymphoproliferation assays unreliable in diagnosis. To investigate the utility and nature of proliferation responses in allergy by comparison of the standard lymphoproliferation with a new dendritic cell (DC) stimulated assay. Monocyte-derived DCs were pulsed with allergens and incubated with autologous T cells for 7 days. DC-stimulated and standard PBMC proliferation responses to 3 common dietary allergens in children with allergy and without atopy were measured by incorporation of tritiated thymidine and reduction of carboxyl fluorescein succinimidyl ester staining. The DC presentation of sensitizing allergens induced significantly higher proliferative responses than PBMC stimulation (P = .04) and greater distinction between normal and allergic responses. DC-induced stimulation indices of children without sensitivity and those with allergy were significantly different with all 3 foods (P < .001). All children with allergy presented with peanut allergy and 12 of 14 (86%) β-lactoglobulin-pulsed DC preparations proliferated more than 3.3-fold above un-pulsed cells, but 8 of 18 children (44%) with ovalbumin egg allergy showed proliferation below this level. The stimulation index of DC tritiated thymidine incorporation correlated closely with carboxyl fluorescein succinimidyl ester reduction (P < .001). Sensitivity of detection of peanut, milk, or egg allergy was 100%, 85.7%, or 55.6% and specificity was 60%, 88.9%, or 86.7%, respectively. DC-stimulated T cells expressed increased levels of CD45 RO and CD25 and most produced interferon-γ. DC-stimulated proliferation correlated with total immunoglobulin E and peanut antigen-stimulated proliferation correlated with peanut specific immunoglobulin E (P = .03). The DC-induced lymphoproliferation had higher sensitivity, specificity, and reproducibility than the standard assay and caused increased memory and activated T-cell proliferation in children with food allergy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Albumin aus Hühnereiweiss, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
β-Lactoglobulin aus Kuhmilch, ≥90% (PAGE), lyophilized powder
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, suitable for (for molecular biology), Non-acetylated, Molecular Biology
Sigma-Aldrich
Trypsin-Inhibitor aus Hühnereiweiss, Type III-O (free of ovoinhibitor)