Merck
  • 4,5-Di-O-Caffeoylquinic Acid from Ligularia fischeri Suppresses Inflammatory Responses Through TRPV1 Activation.
4,5-Di-O-Caffeoylquinic Acid from Ligularia fischeri Suppresses Inflammatory Responses Through TRPV1 Activation.
Phytotherapy research : PTR (2017-08-07)
Yiseul Kim, Jung Tae Kim, Joonwoo Park, Hee Jin Son, Eun-Young Kim, Young Joo Lee, Mee-Ra Rhyu
ZUSAMMENFASSUNG
Ligularia fischeri (Ledeb.) Turcz., a perennial plant native to northeastern Asia, has long been used as folk remedies for the alleviation of inflammatory symptoms. We investigated whether the extract of L. fischeri (LFEx) and caffeoylquinic acid (CQA) derivatives, the pharmacologically active ingredients identified from L. fischeri, regulate inflammation via a transient receptor potential vanilloid 1 (TRPV1)-mediated pathway. Changes in intracellular Ca
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Capsazepine, ≥98% (HPLC), solid
Sigma-Aldrich
3,4-Di-O-caffeoylquinic acid