Merck
N370S-GBA1 mutation causes lysosomal cholesterol accumulation in Parkinson's disease.
Movement disorders : official journal of the Movement Disorder Society (2017-08-06)
Patricia García-Sanz, Lorena Orgaz, Guillermo Bueno-Gil, Isabel Espadas, Eva Rodríguez-Traver, Jaime Kulisevsky, Antonia Gutierrez, José C Dávila, Rosa A González-Polo, José M Fuentes, Pablo Mir, Carlos Vicario, Rosario Moratalla
ZUSAMMENFASSUNG
Heterozygous mutations in the GBA1 gene, which encodes the lysosomal enzyme β-glucocerebrosidase-1, increase the risk of developing Parkinson's disease, although the underlying mechanisms remain unclear. The aim of this study was to explore the impact of the N370S-GBA1 mutation on cellular homeostasis and vulnerability in a patient-specific cellular model of PD. We isolated fibroblasts from 4 PD patients carrying the N370S/wild type GBA1 mutation and 6 controls to study the autophagy-lysosome pathway, endoplasmic reticulum stress, and Golgi apparatus structure by Western blot, immunofluorescence, LysoTracker and Filipin stainings, mRNA analysis, and electron microscopy. We evaluated cell vulnerability by apoptosis, reactive oxygen species and mitochondrial membrane potential with flow cytometry. The N370S mutation produced a significant reduction in β-glucocerebrosidase-1 protein and enzyme activity and β-glucocerebrosidase-1 retention within the endoplasmic reticulum, which interrupted its traffic to the lysosome. This led to endoplasmic reticulum stress activation and triggered unfolded protein response and Golgi apparatus fragmentation. Furthermore, these alterations resulted in autophagosome and p62/SQSTM1 accumulation. This impaired autophagy was a result of dysfunctional lysosomes, indicated by multilamellar body accumulation probably caused by increased cholesterol, enlarged lysosomal mass, and reduced enzyme activity. This phenotype impaired the removal of damaged mitochondria and reactive oxygen species production and enhanced cell death. Our results support a connection between the loss of β-glucocerebrosidase-1 function, cholesterol accumulation, and the disruption of cellular homeostasis in GBA1-PD. Our work reveals new insights into the cellular pathways underlying PD pathogenesis, providing evidence that GBA1-PD shares common features with lipid-storage diseases. © 2017 International Parkinson and Movement Disorder Society.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-LC3B in Kaninchen hergestellte Antikörper, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Pyridiniumchlorchromat, 98%
Sigma-Aldrich
Monoklonaler Anti-β-Tubulin-Antikörper in Maus hergestellte Antikörper, clone TUB 2.1, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Cathepsin D antibody produced in mouse, clone CTD-19, ascites fluid
Sigma-Aldrich
Monoclonal Anti- Cathepsin B in Maus hergestellte Antikörper, clone CB59-4B11, purified from hybridoma cell culture